Learn More
TARGETMOL CHEMICALS INC BAY-678 25MG
SDP
Catalog No. 502581719
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY-678 is a human neutrophil elastase (HNE; IC50 20 nM) inhibitor with oral bioactivity potency and selectivity.BAY-678 is a chemical probe recommended by the Structural Genomics Consortium (SGC). purity: 98%
Catalog No. 50-258-1719 Supplier TARGETMOL CHEMICALS INC Supplier No. T1047325MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY-678 is a human neutrophil elastase (HNE; IC50 20 nM) inhibitor with oral bioactivity potency and selectivity.BAY-678 is a chemical probe recommended by the Structural Genomics Consortium (SGC). purity: 98%