Learn More
Medchemexpress LLC Bay-707 | 2109805-96-9 | 99.6% | 288.34 | 25 MG
SDP
Catalog No. 5000439899
Encompass_Preferred
BAY-707 is a chemical probe and a substrate-competitive, highly potent, and selective inhibitor of MTH1 (NUDT1) with an IC50 of 2.3 nM. It exhibits a favorable pharmacokinetic (PK) profile and is well-tolerated in mice, though it lacks clear in vitro or in vivo anticancer efficacy.
  • Functions as a substrate-competitive inhibitor of MTH1 (NUDT1)
  • Highly potent and selective inhibitor with an IC50 of 2.3 nM
  • Exhibits a favorable pharmacokinetic (PK) profile
  • Well-tolerated in mice
Catalog No. 50-004-39899 Supplier Medchemexpress LLC Supplier No. 25MGHY112081
BAY-707 is a chemical probe and a substrate-competitive, highly potent, and selective inhibitor of MTH1 (NUDT1) with an IC50 of 2.3 nM. It exhibits a favorable pharmacokinetic (PK) profile and is well-tolerated in mice, though it lacks clear in vitro or in vivo anticancer efficacy.
  • Functions as a substrate-competitive inhibitor of MTH1 (NUDT1)
  • Highly potent and selective inhibitor with an IC50 of 2.3 nM
  • Exhibits a favorable pharmacokinetic (PK) profile
  • Well-tolerated in mice