Learn More
Medchemexpress LLC HY-104028 5mg Medchemexpress, BAY 73-6691 CAS:794568-92-6 Purity:>98%
SDP
Catalog No. 501968193
Encompass_Preferred
Medchemexpress, HY-104028 5mg BAY 73-6691 CAS:794568-92-6 BAY 73-6691 is a potent, selective brain penetrant PDE9A inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8193 Supplier Medchemexpress LLC Supplier No. HY1040285MG
Medchemexpress, HY-104028 5mg BAY 73-6691 CAS:794568-92-6 BAY 73-6691 is a potent, selective brain penetrant PDE9A inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.