Learn More
Medchemexpress LLC HY-104028A 5mg Medchemexpress, BAY 73-6691 racemate CAS:794568-90-4 Purity:>98%
SDP
Catalog No. 501968195
Encompass_Preferred
Medchemexpress, HY-104028A 5mg BAY 73-6691 racemate CAS:794568-90-4 BAY 73-6691 racemate is a phosphodiesterase 9 inhibitor extracted from patent WO 2017070293 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8195 Supplier Medchemexpress LLC Supplier No. HY104028A5MG
Medchemexpress, HY-104028A 5mg BAY 73-6691 racemate CAS:794568-90-4 BAY 73-6691 racemate is a phosphodiesterase 9 inhibitor extracted from patent WO 2017070293 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.