Learn More
TARGETMOL CHEMICALS INC BAY-8002 10MG
SDP
Catalog No. 502879778
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BAY-8002 is a potent dual MCT1/2 inhibitor (IC50 values are 3, 8, and 12 nM at rat (C6), human (DLD-1) and mouse (4T1) MCT1 respectively). Purity 99.34%
Catalog No. 50-287-9778 Supplier TARGETMOL CHEMICALS INC Supplier No. T520810MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BAY-8002 is a potent dual MCT1/2 inhibitor (IC50 values are 3, 8, and 12 nM at rat (C6), human (DLD-1) and mouse (4T1) MCT1 respectively). Purity 99.34%