Learn More
Selleck Chemical LLC BAY-8002-5mg
SDP
Supplier:  Selleck Chemical LLC S87475MG
Encompass_Preferred
BAY-8002 is a novel MCT1 inhibitor and potently suppress bidirectional lactate transport. BAY-8002 significantly increases intratumor lactate levels and transiently modulates pyruvate levels.
Catalog No. 50-239-0046