Learn More
Medchemexpress LLC Bay-8002 | 724440-27-1 | 98.8% | 1 ML
SDP
Catalog No. 5000232615
Encompass_Preferred
BAY-8002 is a potent, selective, orally active inhibitor of monocarboxylate transporter 1 (MCT1) used in preclinical oncology and metabolic research. Supplied as a 10 mM solution (1 mL) in DMSO, it is suitable for cell-based assays and mechanistic studies.
  • Potent MCT1 inhibition (IC50 ≈85 nM).
  • Demonstrated antitumor activity in preclinical models.
  • High research-grade purity.
  • Ready-to-use 10 mM DMSO solution for cell assays.
  • Available in solid form for custom formulation.
Catalog No. 50-002-32615 Supplier Medchemexpress LLC Supplier No. HY12231210MM 1ML
BAY-8002 is a potent, selective, orally active inhibitor of monocarboxylate transporter 1 (MCT1) used in preclinical oncology and metabolic research. Supplied as a 10 mM solution (1 mL) in DMSO, it is suitable for cell-based assays and mechanistic studies.
  • Potent MCT1 inhibition (IC50 ≈85 nM).
  • Demonstrated antitumor activity in preclinical models.
  • High research-grade purity.
  • Ready-to-use 10 mM DMSO solution for cell assays.
  • Available in solid form for custom formulation.