Learn More
Medchemexpress LLC BAY-8002 | 724440-27-1 | 98.8% | 415.85 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY122312
Encompass_Preferred
BAY-8002 is a potent, selective, and orally active inhibitor of monocarboxylate transporter 1 (MCT1). It exhibits anti-tumor activity in vivo, significantly inhibiting tumor growth in Raji tumor-bearing mice.
  • Potent and selective MCT1 inhibitor with an IC50 of 85 nM in DLD-1 cells.
  • Excellent selectivity against MCT4 (IC50 >50 μM in EVSA-T cells).
  • Exhibits anti-tumor activity in Raji tumor-bearing mice.
  • Available as a solid, white to off-white appearance.
  • Soluble in DMSO (125 mg/mL).
  • Intended for research use only.
Catalog No. 50-003-65146