Learn More
Medchemexpress LLC Bay-850 | 2099142-76-2 | 98.3% | 100 MG

Supplier: Medchemexpress LLC HY119254100MG
Bay-850 is a small-molecule chemical probe that selectively inhibits ATPase family AAA domain-containing protein 2 (ATAD2). It is characterized as a potent, isoform-selective ATAD2 inhibitor (IC50 166 nM) used in biochemical and cell-based studies to probe ATAD2 function, disrupt ATAD2-chromatin interactions, and induce bromodomain dimerization.
- Potent, isoform-selective ATAD2 inhibition (IC50 166 nM).
- Induces ATAD2 bromodomain dimerization and disrupts ATAD2-chromatin interaction.
- High purity suitable for biochemical assays (98.3% by HPLC).
- Powder form, readily soluble in DMSO for stock solutions.
- Molecular weight 654.24 g/mol; formula C38H44ClN5O3.
- Available with COA and RP-HPLC report for analytical verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.