Learn More
Apexbio Technology LLC BAY 87-2243, 50mg. Cas: 1227158-85-1 MFCD: MFCD28143915. HIF-1 inhibitor,potent and selective
SDP
Catalog No. 501905205
Encompass_Preferred
BAY 87-2243 is a selective inhibitor of HIF-1 induced gene activation [1].In HCT-116 cells, BAY 87-2243 inhibits the hypoxia increased HRE-dependent luciferase expression with IC50 value of 0.7nM and inhibits the expression of HIF target gene CA9 with IC50 value of 2nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5205 Supplier Apexbio Technology LLC Supplier No. B111550
BAY 87-2243 is a selective inhibitor of HIF-1 induced gene activation [1].In HCT-116 cells, BAY 87-2243 inhibits the hypoxia increased HRE-dependent luciferase expression with IC50 value of 0.7nM and inhibits the expression of HIF target gene CA9 with IC50 value of 2nM. Other sizes are available. Please inqury us for quote.