Learn More
Medchemexpress LLC HY-100017 10mg Medchemexpress, BAY-876 CAS:1799753-84-6 Purity:>98%
SDP
Catalog No. 502028600
Encompass_Preferred
Medchemexpress, HY-100017 10mg BAY-876 CAS:1799753-84-6 BAY-876 is an orally active and selective glucose transporter 1 (GLUT1) inhibitor with an IC50 of 2 nM. BAY-876 is >130-fold more selective for GLUT1 than GLUT2, GLUT3, and GLUT4. BAY-876 is also a potent blocker of glycolytic metabolism and ovarian cancer growth[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8600 Supplier Medchemexpress LLC Supplier No. HY10001710MG
Medchemexpress, HY-100017 10mg BAY-876 CAS:1799753-84-6 BAY-876 is an orally active and selective glucose transporter 1 (GLUT1) inhibitor with an IC50 of 2 nM. BAY-876 is >130-fold more selective for GLUT1 than GLUT2, GLUT3, and GLUT4. BAY-876 is also a potent blocker of glycolytic metabolism and ovarian cancer growth[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.