Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BAY-899 | 2471967-92-5 | 99.9% | 100 MG
SDP

Catalog No. 5000338971
Encompass_Preferred

BAY-899 is a chemical probe and an orally active, selective luteinizing hormone receptor (LH-R) antagonist. It is characterized by IC50s of 185 nM for human LH and 46 nM for rat LH, and has shown efficacy in reducing sex hormone levels in vivo. This product is intended for research use only.

  • High purity of 99.93%
  • Selective luteinizing hormone receptor (LH-R) antagonist
  • Reduces sex hormone levels
  • Potent inhibition with IC50s of 185 nM for human LH and 46 nM for rat LH
  • Available as a white to off-white solid
  • Stable as powder for up to 3 years at -20°C or 2 years at 4°C

Catalog No. 50-003-38971 Supplier Medchemexpress LLC Supplier No. HY130248100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BAY-899 is a chemical probe and an orally active, selective luteinizing hormone receptor (LH-R) antagonist. It is characterized by IC50s of 185 nM for human LH and 46 nM for rat LH, and has shown efficacy in reducing sex hormone levels in vivo. This product is intended for research use only.

  • High purity of 99.93%
  • Selective luteinizing hormone receptor (LH-R) antagonist
  • Reduces sex hormone levels
  • Potent inhibition with IC50s of 185 nM for human LH and 46 nM for rat LH
  • Available as a white to off-white solid
  • Stable as powder for up to 3 years at -20°C or 2 years at 4°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.