Learn More
Medchemexpress LLC Bay-899 | 2471967-92-5 | 99.9% | C25H19F2N5O2 | 10MG
SDP
Catalog No. 5000259604
Encompass_Preferred
BAY-899 is a tetrahydro-1,6-naphthyridine-based small-molecule antagonist of the luteinizing hormone receptor (LH-R), used as a research probe to modulate LH-R signaling and reduce sex hormone levels in vitro and in vivo.
  • Orally active small molecule with demonstrated in vivo activity.
  • Selective LH-R antagonism with IC50s: human 185 nM, rat 46 nM.
  • High chemical purity (99.9%) suitable for biochemical assays.
  • Molecular formula C25H19F2N5O2; molecular weight 459.45 g·mol⁻¹.
  • Supplied as a solid for solution preparation and dosing studies.
Catalog No. 50-002-59604 Supplier Medchemexpress LLC Supplier No. HY13024810MG
BAY-899 is a tetrahydro-1,6-naphthyridine-based small-molecule antagonist of the luteinizing hormone receptor (LH-R), used as a research probe to modulate LH-R signaling and reduce sex hormone levels in vitro and in vivo.
  • Orally active small molecule with demonstrated in vivo activity.
  • Selective LH-R antagonism with IC50s: human 185 nM, rat 46 nM.
  • High chemical purity (99.9%) suitable for biochemical assays.
  • Molecular formula C25H19F2N5O2; molecular weight 459.45 g·mol⁻¹.
  • Supplied as a solid for solution preparation and dosing studies.