Learn More
Selleck Chemical LLC BAY-985 S8935-5MG
SDP
Catalog No. 502173445
Encompass_Preferred
BAY-985 is a potent and highly selective TBK1/IKK inhibitor BAY-985 shows high potency toward TBK1 (IC50 2 nM low ATP assay 30 nM high ATP assay) and IKK (IC50 2 nM) as well as high potency in the mechanistic pIRF3 assay (IC50 74 nM) and an antiproliferative effect on SK-MEL-2 cells (IC50 900 nM)
Catalog No. 50-217-3445 Supplier Selleck Chemical LLC Supplier No. S89355MG
BAY-985 is a potent and highly selective TBK1/IKK inhibitor BAY-985 shows high potency toward TBK1 (IC50 2 nM low ATP assay 30 nM high ATP assay) and IKK (IC50 2 nM) as well as high potency in the mechanistic pIRF3 assay (IC50 74 nM) and an antiproliferative effect on SK-MEL-2 cells (IC50 900 nM)