Learn More
Medchemexpress LLC HY-10588 5mg , Bay K 8644 CAS:71145-03-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105885MG
Encompass_Preferred
HY-10588 5mg Bay K 8644 CAS: 71145-03-4 Bay K 8644 ((±)-Bay K 8644) is a racemate consisting of two isomers (R)-(+)-Bay-K-8644 and (S)-(-)-Bay-K-8644. Bay K 8644 is a L-type Ca2+ channel agonist with an EC50 of 17.3 nM. Bay K 8644 increases Ca2+ influx through sarcolemmal Ca2+ channels by increasing the open time of the channel. Bay K 8644 has vasoconstrictive effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1634