Learn More
Medchemexpress LLC (±)-Bay K 8644 | 71145-03-4 | MFCD00036697 | 100.0% | 356.30 g·mol⁻¹ | C16H15F3N2O4 | 10 MG
SDP
Catalog No. 5000243709
Encompass_Preferred
Bay K 8644 ((±)-Bay K 8644) is a racemic L-type calcium channel agonist used as a research reagent to increase Ca2+ influx by prolonging the open time of sarcolemmal L-type Ca2+ channels. It is applied in pharmacology and electrophysiology studies and exhibits vasoconstrictive activity.
  • Acts as an L-type Ca2+ channel agonist.
  • Racemic mixture comprising two enantiomers.
  • Potent activity with reported EC50 of 17.3 nM.
  • High purity suitable for research (99.96%).
  • Molecular weight 356.30 g·mol⁻¹; formula C16H15F3N2O4.
  • Available in small milligram pack sizes for laboratory use.
Catalog No. 50-002-43709 Supplier Medchemexpress LLC Supplier No. HY1058810MG
Bay K 8644 ((±)-Bay K 8644) is a racemic L-type calcium channel agonist used as a research reagent to increase Ca2+ influx by prolonging the open time of sarcolemmal L-type Ca2+ channels. It is applied in pharmacology and electrophysiology studies and exhibits vasoconstrictive activity.
  • Acts as an L-type Ca2+ channel agonist.
  • Racemic mixture comprising two enantiomers.
  • Potent activity with reported EC50 of 17.3 nM.
  • High purity suitable for research (99.96%).
  • Molecular weight 356.30 g·mol⁻¹; formula C16H15F3N2O4.
  • Available in small milligram pack sizes for laboratory use.