Learn More
Medchemexpress LLC BAY-Y 3118 | 151213-16-0 | 405.85 | 1 ML
SDP
Catalog No. 5000448816
Encompass_Preferred
BAY-Y 3118 is a quinolone antibacterial agent with a broad antibacterial spectrum in vitro, showing high activity against gram-positive cocci and anaerobes, and moderate activity against Enterobacteriaceae and Pseudomonas aeruginosa. It is suitable for the research of infectious diseases.
  • Broad antibacterial spectrum in vitro
  • Highly active against gram-positive cocci and anaerobes
  • Moderately active against Enterobacteriaceae and Pseudomonas aeruginosa
  • Reaches high intracellular concentrations and remains active intracellularly
  • Effectively eliminates *L. monocytogenes* from infected cells and organs
  • Rapidly bactericidal in vitro
Catalog No. 50-004-48816 Supplier Medchemexpress LLC Supplier No. HYU0009210MM/1ML
BAY-Y 3118 is a quinolone antibacterial agent with a broad antibacterial spectrum in vitro, showing high activity against gram-positive cocci and anaerobes, and moderate activity against Enterobacteriaceae and Pseudomonas aeruginosa. It is suitable for the research of infectious diseases.
  • Broad antibacterial spectrum in vitro
  • Highly active against gram-positive cocci and anaerobes
  • Moderately active against Enterobacteriaceae and Pseudomonas aeruginosa
  • Reaches high intracellular concentrations and remains active intracellularly
  • Effectively eliminates *L. monocytogenes* from infected cells and organs
  • Rapidly bactericidal in vitro