Learn More
Medchemexpress LLC BAY-Y 3118 | 151213-16-0 | MFCD31557930 | 405.85 | 25 MG
SDP
Catalog No. 5000448243
Encompass_Preferred
BAY-Y 3118 is a quinolone antibacterial agent with a broad antibacterial spectrum in vitro. It demonstrates high activity against gram-positive cocci and anaerobes, and moderate activity against Enterobacteriaceae and *Pseudomonas aeruginosa*. It can be utilized in the research of infectious diseases.
  • Potent against *Haemophilus influenzae*, *Moraxella catarrhalis*, *Acinetobacter baumannii*, *Xanthomonas maltophilia*, gram-positive cocci, and anaerobes.
  • Achieves high intracellular concentrations within human polymorphonuclear leukocytes (PMNs).
  • Highly susceptible to *Listeria monocytogenes* and other *Listeria spp*.
  • Rapid bactericidal effects in vitro and a postantibiotic effect lasting 3 hours.
  • Can eliminate *L. monocytogenes* from infected L929 cells.
  • Cures immunocompetent mice treated with 4 mg every 12 hours.
  • Eliminates *L. monocytogenes* from livers and spleens of chronically infected nude mice.
Catalog No. 50-004-48243 Supplier Medchemexpress LLC Supplier No. 25MGHYU00092
BAY-Y 3118 is a quinolone antibacterial agent with a broad antibacterial spectrum in vitro. It demonstrates high activity against gram-positive cocci and anaerobes, and moderate activity against Enterobacteriaceae and *Pseudomonas aeruginosa*. It can be utilized in the research of infectious diseases.
  • Potent against *Haemophilus influenzae*, *Moraxella catarrhalis*, *Acinetobacter baumannii*, *Xanthomonas maltophilia*, gram-positive cocci, and anaerobes.
  • Achieves high intracellular concentrations within human polymorphonuclear leukocytes (PMNs).
  • Highly susceptible to *Listeria monocytogenes* and other *Listeria spp*.
  • Rapid bactericidal effects in vitro and a postantibiotic effect lasting 3 hours.
  • Can eliminate *L. monocytogenes* from infected L929 cells.
  • Cures immunocompetent mice treated with 4 mg every 12 hours.
  • Eliminates *L. monocytogenes* from livers and spleens of chronically infected nude mice.