Learn More
Medchemexpress LLC HY-100018 2mg Medchemexpress, BAY1125976 CAS:1402608-02-9 Purity:>98%
SDP
Catalog No. 501931692
Encompass_Preferred
Medchemexpress, HY-100018 2mg BAY1125976 CAS:1402608-02-9 BAY1125976 is a selective allosteric Akt1/Akt2 inhibitor; inhibits Akt1 and Akt2 activity with IC 50 values of 5.2 nM and 18 nM at 10 μM ATP, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1692 Supplier Medchemexpress LLC Supplier No. HY1000182MG
Add to Cart
Add to Cart
Medchemexpress, HY-100018 2mg BAY1125976 CAS:1402608-02-9 BAY1125976 is a selective allosteric Akt1/Akt2 inhibitor; inhibits Akt1 and Akt2 activity with IC 50 values of 5.2 nM and 18 nM at 10 μM ATP, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.