Learn More
Medchemexpress LLC HY-112316 10mg Medchemexpress, BAY1238097 CAS:1564268-08-1 Purity:>98%
SDP
Catalog No. 501969080
Encompass_Preferred
Medchemexpress, HY-112316 10mg BAY1238097 CAS:1564268-08-1 BAY1238097 is a potent and selective inhibitor of BET binding to histones and has strong anti-proliferative activity in different AML (acute myeloid leukemia) and MM (multiple myeloma) models through down-regulation of c-Myc levels and its downstream transcriptome (IC50 <100 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9080 Supplier Medchemexpress LLC Supplier No. HY11231610MG
Medchemexpress, HY-112316 10mg BAY1238097 CAS:1564268-08-1 BAY1238097 is a potent and selective inhibitor of BET binding to histones and has strong anti-proliferative activity in different AML (acute myeloid leukemia) and MM (multiple myeloma) models through down-regulation of c-Myc levels and its downstream transcriptome (IC50 <100 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.