Learn More
Siemens Healthineers Bayer™ Calibration and Slope Cylinder Gases
Catalog No. 09529226 Shop All Siemens Healthineers Products
Change view
Click to view available options
Type:
Calibration Gas
Slope Gas
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-529-226 Calibration Gas
09-529-227 Slope Gas
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-529-226 Supplier Siemens Healthineers Supplier No. 10311037
Add to Cart
Add to Cart
For blood gas systems requiring cylinders
  • Choice of calibration or slope gas for use with any blood gas system
  • Packaged in disposable E-D cylinders
  • Each type of gas has individual physical properties
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Blood gas system
Type Calibration Gas
Includes 5% Carbon Dioxide, 12% Oxygen, balance Nitrogen
Quantity Each
WARNING: Reproductive Harm - www.P65Warnings.ca.gov