Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Bayer™ Model 600 Series Electrolyte Analyzers: Electrodes

Catalog No. 09529264 Shop All Siemens Healthineers Products
Change view
Click to view available options
Type:
Ca+ Electrode
Reference Electrode Cassette, 4 M KCl Solution
Reference Electrode Fill Solution KCl
Reference Electrode, Body/KCl, 4 M KCl Solution and Inner Element2
pH Electrode2
For Use With (Equipment):
Bayer Electrolyte Analyzers: 614, 634
Bayer Electrolyte Analyzers: 614, 634, 644, 654
Bayer Electrolyte Analyzers: 614, 654
Bayer Electrolyte Analyzers: 644, 654
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
09-529-264 Reference Electrode Fill Solution KCl Bayer Electrolyte Analyzers: 644, 654
09-529-253 Ca+ Electrode Bayer Electrolyte Analyzers: 614, 634
09-529-252 pH Electrode2 Bayer Electrolyte Analyzers: 614, 634
09-529-254 Reference Electrode, Body/KCl, 4 M KCl Solution and Inner Element2 Bayer Electrolyte Analyzers: 614, 654
09-529-263 Reference Electrode Cassette, 4 M KCl Solution Bayer Electrolyte Analyzers: 614, 634, 644, 654
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 09-529-264 Supplier Siemens Healthineers Supplier No. 10311081
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Electrodes for use with Bayer™ 600 Series Electrolyte Analyzers.

For use with Bayer Electrolyte Analyzer models 644 and 654.

1 Warranted for 90 days from date of shipment.

2 Warranted for 180 days from date of shipment.

3 Contains 4 x 370mL calibrating solution; 4 x 75mL slope standardizing solution; 4 deproteinizing/conditioning packs.

TRUSTED_SUSTAINABILITY

Specifications

Type Reference Electrode Fill Solution KCl
For Use With (Application) Electrolyte analysis
For Use With (Equipment) Bayer Electrolyte Analyzers: 644, 654
Capacity (Metric) 5 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.