Learn More
Siemens Healthineers Bayer™ RapidLab™ Deproteinizing Kit
Catalog No. 09529267A Shop All Siemens Healthineers Products
Change view
Click to view available options
For Use With (Equipment):
RapidLab 800 Series, M248, M238, M654, M348, M614, M1200 Analyzers
Catalog No. For Use With (Equipment)
09-529-267A RapidLab 800 Series, M248, M238, M654, M348, M614, M1200 Analyzers
1 options
This item is not returnable. View return policy
Catalog No. 09-529-267A Supplier Siemens Healthineers Supplier No. 105610
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
For use with Bayer RapidLab 238, 248, 348 Blood Gas Analyzer
  • Manufacturer recommends use weekly
  • Takes five minutes
TRUSTED_SUSTAINABILITY
Specifications
Type Deproteinizing Kit
For Use With (Equipment) RapidLab 800 Series, M248, M238, M654, M348, M614, M1200 Analyzers