Learn More
Ascensia Diabetes Care Urinalysis Reagent Test Strips
Description
- Ready to use and entirely disposable
- Dip and read visually for results
- Some strips can be read instrumentally on Clinitek™ 50, Clinitek 100, Clinitek 200, Clinitek 200+ and Clinitek 500 Urine Chemistry analyzers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Clia Complexity | Waived |
| Detectable Analytes | Glucose, Ketone |
| Disposable | Single-use |
| For Use With (Application) | Urinalysis |
| Quantity | 100 Pack |