Learn More
Cayman Chemical Bayogenin, Purity: ≥90%, CAS: 6989-24-8, 10 mg
SDP
Catalog No. 502910844
Encompass_Preferred
A triterpene with enzyme inhibitory activity; inhibits rabbit muscle glycogen phosphorylase (IC50 = 68 µM)
Catalog No. 50-291-0844 Supplier Cayman Chemical Supplier No. 3582210 MG
A triterpene with enzyme inhibitory activity; inhibits rabbit muscle glycogen phosphorylase (IC50 = 68 µM)