Learn More
Hach Company Baysilone silicone grease, 35 g tube - lubricant for O-rings and seals (Hach LZV678)
Catalog No. NC3842295 Shop All Hach Company Products
Encompass
Baysilone silicone grease is a silicone-based lubricant supplied in a 35 g tube intended for maintenance of elastomeric components. It provides stable, long-lasting lubrication to reduce wear and preserve seal performance in valves, fittings, and other movable parts used with analytical equipment.
  • 35 g tube packaging.
  • Silicone-based grease for stable lubrication.
  • Suitable for lubricating O-rings and seals.
  • Helps reduce friction and wear on elastomeric parts.
  • Easy-to-apply consistency for small maintenance tasks.
Catalog No. NC3842295 Supplier Hach Company Supplier No. LZV678
Baysilone silicone grease is a silicone-based lubricant supplied in a 35 g tube intended for maintenance of elastomeric components. It provides stable, long-lasting lubrication to reduce wear and preserve seal performance in valves, fittings, and other movable parts used with analytical equipment.
  • 35 g tube packaging.
  • Silicone-based grease for stable lubrication.
  • Suitable for lubricating O-rings and seals.
  • Helps reduce friction and wear on elastomeric parts.
  • Easy-to-apply consistency for small maintenance tasks.