Learn More
Aviva Systems Biology Corp BAZ1A Antibody - middle region (ARP32848_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32848_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAZ1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BAZ1A. Alternate names: ACF1, DKFZP586E0518, FLJ14383, WALp1, WCRF180, hACF1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54793