Learn More
Eurofins Discoverx BAZ2-ICR, 10 mg. (92-1308)
Supplier:  Eurofins Discoverx 921308
Encompass
BAZ2-ICR or 4-[4-(1-Methyl-1H-pyrazole-4-yl)-1-[2-(1-methyl-1H-pyrazol-4-yl)ethyl]-1H-imidazol-5-yl]benzonitrile, is a selective BAZ2 bromodomain inhibitor that exhibits 15-fold selectivity for the BAZ2 bromodomain over the CERC2 bromodomain and >100-fold selectivity over a range of other bromodomains. Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9822