Learn More
Medchemexpress LLC HY-19336 5mg Medchemexpress, BAZ2-ICR CAS:1665195-94-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY193365MG
Encompass_Preferred
Medchemexpress, HY-19336 5mg BAZ2-ICR CAS:1665195-94-7 BAZ2-ICR is a potent, selective, cell active and orally active BAZ2A/B bromodomains inhibitor with IC50s of 130 nM and 180 nM, and Kds of 109 nM and 170 nM, respectively. BAZ2-ICR shows 10-15-fold selectivity for binding BAZ2A/B over CECR2 and >100-fold selectivity over all other bromodomains. BAZ2-ICR is an epigenetic chemical probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0023