Learn More
Cayman Chemical BazedoxIfen-d4 500ug
SDP
Catalog No. 502964132
Encompass_Preferred
An internal standard for the quantification of bazedoxifene by GC- or LC-MS
Catalog No. 50-296-4132 Supplier Cayman Chemical Supplier No. 22554500 UG
An internal standard for the quantification of bazedoxifene by GC- or LC-MS