Learn More
Cayman Chemical Bazedoxifene (acetate), Purity: ≥98%, CAS: 198481-33-3, 50 mg
SDP
Catalog No. 502972021
Encompass_Preferred
An indole-based SERM that binds to both ERα (IC50 = 26 nM) and ERβ (IC50 = 99 nM); antagonizes 17β-estradiol-dependent and hormone-independent growth of breast cancer cell proliferation (80% reduction with 10 nM) in a manner related to cell cycle arrest and downregulation of cyclin D1 and ERα
Catalog No. 50-297-2021 Supplier Cayman Chemical Supplier No. 1500550 MG
An indole-based SERM that binds to both ERα (IC50 = 26 nM) and ERβ (IC50 = 99 nM); antagonizes 17β-estradiol-dependent and hormone-independent growth of breast cancer cell proliferation (80% reduction with 10 nM) in a manner related to cell cycle arrest and downregulation of cyclin D1 and ERα