Learn More
Medchemexpress LLC HY-A0031 10mg , Bazedoxifene CAS:198481-32-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA003110MG
Encompass_Preferred
HY-A0031 10mg Bazedoxifene CAS: 198481-32-2 Bazedoxifene (TSE-424) is an oral, BBB-penetrant nonsteroidal selective estrogen receptor modulator (SERM), with IC50s of 23 nM and 99 nM for ERα and ERβ, respectively. Bazedoxifene can be used for the research of osteoporosis. Bazedoxifene also acts as an inhibitor of IL-6/GP130 protein-protein interactions and can be used for the research of pancreatic cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1516