Learn More
Medchemexpress LLC Bazedoxifene | 198481-32-2 | 99.1% | C30H34N2O3 | 100 MG
SDP
Catalog No. 5000432346
Encompass_Preferred
Bazedoxifene is an oral, BBB-penetrant nonsteroidal selective estrogen receptor modulator (SERM). It is useful for osteoporosis research and inhibits IL-6/GP130 protein-protein interactions, making it suitable for pancreatic cancer research.
  • Oral, BBB-penetrant nonsteroidal selective estrogen receptor modulator
  • Useful in osteoporosis research
  • Inhibits IL-6/GP130 protein-protein interactions
  • Can be used for pancreatic cancer research
  • IC50s of 23 nM for ERα and 99 nM for ERβ
Catalog No. 50-004-32346 Supplier Medchemexpress LLC Supplier No. HYA0031100MG
Bazedoxifene is an oral, BBB-penetrant nonsteroidal selective estrogen receptor modulator (SERM). It is useful for osteoporosis research and inhibits IL-6/GP130 protein-protein interactions, making it suitable for pancreatic cancer research.
  • Oral, BBB-penetrant nonsteroidal selective estrogen receptor modulator
  • Useful in osteoporosis research
  • Inhibits IL-6/GP130 protein-protein interactions
  • Can be used for pancreatic cancer research
  • IC50s of 23 nM for ERα and 99 nM for ERβ