Learn More
Medchemexpress LLC Bazedoxifene (acetate) | 198481-33-3 | 99.88% | C32H38N2O5 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYA0036500MG
Encompass_Preferred
Bazedoxifene acetate (TSE-424 acetate) is an oral, nonsteroidal selective estrogen receptor modulator (SERM). It has IC50s of 23 nM for ERα and 99 nM for ERβ. This compound is used in the research of osteoporosis. Additionally, Bazedoxifene acetate acts as an inhibitor of IL-6/GP130 protein-protein interactions and is utilized in research for pancreatic cancer.
  • Oral, nonsteroidal selective estrogen receptor modulator (SERM)
  • Exhibits IC50s of 23 nM for ERα and 99 nM for ERβ
  • Used in osteoporosis and pancreatic cancer research
  • Inhibits IL-6/GP130 protein-protein interactions and STAT3 phosphorylation
  • Induces apoptosis and blocks cell migration in pancreatic cancer cells in vitro
  • Inhibits Capan-1 tumor growth in a mouse model in vivo
Catalog No. 50-004-32669