Learn More
Medchemexpress LLC Bazedoxifene acetate | 198481-33-3 | 99.9% | 10 MG
SDP
Catalog No. 5000299254
Encompass_Preferred
Bazedoxifene acetate is an orally active, nonsteroidal selective estrogen receptor modulator (SERM) supplied as a solid research reagent for studies of estrogen receptor signaling, osteoporosis, and cancer. It shows activity against ERα and ERβ and is used in biochemical and cell-based assays.
Catalog No. 50-002-99254 Supplier Medchemexpress LLC Supplier No. HYA0036B10MG
Bazedoxifene acetate is an orally active, nonsteroidal selective estrogen receptor modulator (SERM) supplied as a solid research reagent for studies of estrogen receptor signaling, osteoporosis, and cancer. It shows activity against ERα and ERβ and is used in biochemical and cell-based assays.