Learn More
Medchemexpress LLC Bazedoxifene acetate | 198481-33-3 | 99.9% | 530.65 | C32H38N2O5 | 10 MG
SDP
Catalog No. 5000297909
Encompass_Preferred
Bazedoxifene acetate (D-crystal form) is an orally active, nonsteroidal selective estrogen receptor modulator (SERM) provided for research use in biological and pharmacological studies. It is supplied as a white to off-white solid and is used to probe estrogen receptor signaling and related pathways in vitro and in vivo.
  • D-crystal form with high listed purity (≈99.9%).
  • Nonsteroidal SERM active at ERα and ERβ for receptor modulation studies.
  • Supplied in small research quantities suitable for dose-response experiments.
  • White to off-white solid suitable for analytical and formulation work.
  • Store sealed away from moisture and light; follow low-temperature storage for solutions.
Catalog No. 50-002-97909 Supplier Medchemexpress LLC Supplier No. HYA0036D10MG
Bazedoxifene acetate (D-crystal form) is an orally active, nonsteroidal selective estrogen receptor modulator (SERM) provided for research use in biological and pharmacological studies. It is supplied as a white to off-white solid and is used to probe estrogen receptor signaling and related pathways in vitro and in vivo.
  • D-crystal form with high listed purity (≈99.9%).
  • Nonsteroidal SERM active at ERα and ERβ for receptor modulation studies.
  • Supplied in small research quantities suitable for dose-response experiments.
  • White to off-white solid suitable for analytical and formulation work.
  • Store sealed away from moisture and light; follow low-temperature storage for solutions.