Learn More
Medchemexpress LLC Bazedoxifene (acetate) | 198481-33-3 | 99.9% | C32H38N2O5 | 200 MG
SDP
Catalog No. 5000433506
Encompass_Preferred
Bazedoxifene acetate (TSE-424 acetate) is an oral, nonsteroidal selective estrogen receptor modulator (SERM), with IC50s of 23 nM and 99 nM for ERα and ERβ, respectively. It can be used for the research of osteoporosis. Bazedoxifene acetate also acts as an inhibitor of IL-6/GP130 protein-protein interactions and can be used for the research of pancreatic cancer.
  • Oral, nonsteroidal selective estrogen receptor modulator (SERM)
  • IC50s of 23 nM and 99 nM for ERα and ERβ
  • Useful for the research of osteoporosis
  • Acts as an inhibitor of IL-6/GP130 protein-protein interactions
  • Useful for the research of pancreatic cancer
Catalog No. 50-004-33506 Supplier Medchemexpress LLC Supplier No. HYA0036200MG
Bazedoxifene acetate (TSE-424 acetate) is an oral, nonsteroidal selective estrogen receptor modulator (SERM), with IC50s of 23 nM and 99 nM for ERα and ERβ, respectively. It can be used for the research of osteoporosis. Bazedoxifene acetate also acts as an inhibitor of IL-6/GP130 protein-protein interactions and can be used for the research of pancreatic cancer.
  • Oral, nonsteroidal selective estrogen receptor modulator (SERM)
  • IC50s of 23 nM and 99 nM for ERα and ERβ
  • Useful for the research of osteoporosis
  • Acts as an inhibitor of IL-6/GP130 protein-protein interactions
  • Useful for the research of pancreatic cancer