Learn More
Medchemexpress LLC Bazedoxifene (acetate) | 198481-33-3 | 99.9% | C32H38N2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYA0036B
Encompass_Preferred
Bazedoxifene (acetate)-B is the B-crystal form compound of Bazedoxifene acetate. Bazedoxifene acetate is an orally active, nonsteroidal selective estrogen receptor modulator (SERM), with IC50s of 23 nM for ERα and 99 nM for ERβ.
  • B-crystal form compound of bazedoxifene acetate
  • Orally active nonsteroidal selective estrogen receptor modulator
  • IC50s of 23 nM for ERα and 99 nM for ERβ
  • Solid appearance
  • For research use only
  • Stable under recommended storage conditions
Catalog No. 50-003-58402