Learn More
Medchemexpress LLC Bazedoxifene (acetate) | 198481-33-3 | 99.9% | C32H38N2O5 | 25 MG
SDP
Catalog No. 5000359228
Encompass_Preferred
Bazedoxifene acetate is an oral, nonsteroidal selective estrogen receptor modulator (SERM) that acts as an inhibitor of IL-6/GP130 protein-protein interactions. It is used in research for osteoporosis and pancreatic cancer.
  • Small molecular GP130 inhibitor
  • Inhibits STAT3 phosphorylation induced by IL-6 and IL-11
  • Induces apoptosis in human pancreatic cancer cells
  • Inhibits STAT3 nuclear translocation induced by IL-6
  • Blocks cell migration in pancreatic cancer cells by inhibiting GP130
  • Inhibits Capan-1 tumor growth in mouse models
Catalog No. 50-003-59228 Supplier Medchemexpress LLC Supplier No. 25MGHYA0036
Bazedoxifene acetate is an oral, nonsteroidal selective estrogen receptor modulator (SERM) that acts as an inhibitor of IL-6/GP130 protein-protein interactions. It is used in research for osteoporosis and pancreatic cancer.
  • Small molecular GP130 inhibitor
  • Inhibits STAT3 phosphorylation induced by IL-6 and IL-11
  • Induces apoptosis in human pancreatic cancer cells
  • Inhibits STAT3 nuclear translocation induced by IL-6
  • Blocks cell migration in pancreatic cancer cells by inhibiting GP130
  • Inhibits Capan-1 tumor growth in mouse models