Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bazedoxifene acetate | 198481-33-3 | MFCD09260074 | 99% | C32H38N2O5 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYA0036B100MG

Encompass_Preferred

Bazedoxifene acetate, also known as TSE-424 (acetate)-B, is the B-crystal form compound of Bazedoxifene acetate. It functions as an orally active, nonsteroidal selective estrogen receptor modulator (SERM). This product is intended for research use only.

  • Orally active, nonsteroidal selective estrogen receptor modulator (SERM)
  • IC50s of 23 nM for ERα and 99 nM for ERβ
  • Ships at room temperature
  • Store at 4°C, sealed, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 50 mg/mL

Catalog No. 50-004-32670


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.