Learn More
Medchemexpress LLC Bazedoxifene (acetate)-B | 198481-33-3 | C32H38N2O5 | 5 MG
SDP
Catalog No. 5000433508
Encompass_Preferred
Bazedoxifene (acetate)-B (TSE-424 (acetate)-B) is the B-crystal form compound of Bazedoxifene acetate. It is an orally active, nonsteroidal selective estrogen receptor modulator (SERM), with IC50s of 23 nM and 99 nM for ERα and ERβ, respectively. It is intended for research use only.
  • Orally active
  • Nonsteroidal selective estrogen receptor modulator (SERM)
  • IC50s of 23 nM and 99 nM for ERα and ERβ
  • Intended for research use only
Catalog No. 50-004-33508 Supplier Medchemexpress LLC Supplier No. HYA0036B5MG
Bazedoxifene (acetate)-B (TSE-424 (acetate)-B) is the B-crystal form compound of Bazedoxifene acetate. It is an orally active, nonsteroidal selective estrogen receptor modulator (SERM), with IC50s of 23 nM and 99 nM for ERα and ERβ, respectively. It is intended for research use only.
  • Orally active
  • Nonsteroidal selective estrogen receptor modulator (SERM)
  • IC50s of 23 nM and 99 nM for ERα and ERβ
  • Intended for research use only