Learn More
Medchemexpress LLC Bazedoxifene (acetate)-B (TSE-424 (acetate)-B) | 198481-33-3 | C32H38N2O5 | 50 MG
SDP
Catalog No. 5000432671
Encompass_Preferred
Bazedoxifene (acetate)-B is the B-crystal form compound of Bazedoxifene acetate. Bazedoxifene acetate is an orally active, nonsteroidal selective estrogen receptor modulator (SERM).
  • It is an orally active compound.
  • It functions as a nonsteroidal selective estrogen receptor modulator.
  • It exhibits an IC50 of 23 nM for ERα.
  • It exhibits an IC50 of 99 nM for ERβ.
  • It is a B-crystal form compound.
Catalog No. 50-004-32671 Supplier Medchemexpress LLC Supplier No. HYA0036B50MG
Bazedoxifene (acetate)-B is the B-crystal form compound of Bazedoxifene acetate. Bazedoxifene acetate is an orally active, nonsteroidal selective estrogen receptor modulator (SERM).
  • It is an orally active compound.
  • It functions as a nonsteroidal selective estrogen receptor modulator.
  • It exhibits an IC50 of 23 nM for ERα.
  • It exhibits an IC50 of 99 nM for ERβ.
  • It is a B-crystal form compound.