Learn More
Medchemexpress LLC Bazedoxifene (acetate)-D | 198481-33-3 | C32H38N2O5 | 5 MG
SDP
Catalog No. 5000432330
Encompass_Preferred
Bazedoxifene (acetate)-D is the D-crystal form compound of Bazedoxifene acetate, an orally active, nonsteroidal selective estrogen receptor modulator (SERM). It targets the Estrogen Receptor/ERR pathway, specifically the Vitamin D Related/Nuclear Receptor, with IC50s of 23 nM for ERα and 99 nM for ERβ.
  • D-crystal form compound
  • Orally active, nonsteroidal selective estrogen receptor modulator (SERM)
  • Targets estrogen receptor/ERR
  • Part of vitamin D related/nuclear receptor pathway
  • IC50s of 23 nM for ERα and 99 nM for ERβ
Catalog No. 50-004-32330 Supplier Medchemexpress LLC Supplier No. HYA0036D5MG
Bazedoxifene (acetate)-D is the D-crystal form compound of Bazedoxifene acetate, an orally active, nonsteroidal selective estrogen receptor modulator (SERM). It targets the Estrogen Receptor/ERR pathway, specifically the Vitamin D Related/Nuclear Receptor, with IC50s of 23 nM for ERα and 99 nM for ERβ.
  • D-crystal form compound
  • Orally active, nonsteroidal selective estrogen receptor modulator (SERM)
  • Targets estrogen receptor/ERR
  • Part of vitamin D related/nuclear receptor pathway
  • IC50s of 23 nM for ERα and 99 nM for ERβ