Learn More
Medchemexpress LLC Bazedoxifene (acetate)-D | 198481-33-3 | MFCD09260074 | C32H38N2O5 | 100 MG
SDP
Catalog No. 5000433089
Encompass_Preferred
Bazedoxifene (acetate)-D is a solid compound with a molecular formula of C32H38N2O5 and a molecular weight of 530.65. It is classified as a selective estrogen receptor modulator and a bone density conservation agent. This compound helps to inhibit bone resorption and favor bone mineralization, making it suitable for addressing conditions like osteoporosis and promoting bone fracture healing. It is an FDA-approved, prescription-only drug administered orally.
  • Functions as a selective estrogen receptor modulator
  • Acts as a bone density conservation agent
  • Inhibits bone resorption
  • Promotes bone mineralization and regeneration
  • Utilized in treating osteoporosis and healing bone fractures
  • FDA-approved, available by prescription only
  • Administered via oral route
Catalog No. 50-004-33089 Supplier Medchemexpress LLC Supplier No. HYA0036D100MG
Bazedoxifene (acetate)-D is a solid compound with a molecular formula of C32H38N2O5 and a molecular weight of 530.65. It is classified as a selective estrogen receptor modulator and a bone density conservation agent. This compound helps to inhibit bone resorption and favor bone mineralization, making it suitable for addressing conditions like osteoporosis and promoting bone fracture healing. It is an FDA-approved, prescription-only drug administered orally.
  • Functions as a selective estrogen receptor modulator
  • Acts as a bone density conservation agent
  • Inhibits bone resorption
  • Promotes bone mineralization and regeneration
  • Utilized in treating osteoporosis and healing bone fractures
  • FDA-approved, available by prescription only
  • Administered via oral route