Learn More
Cayman Chemical BB-22 3-carboxyindole metabol
SDP
Catalog No. 502970132
Encompass_Preferred
An expected metabolite of BB-22 that would be detectable both in serum and urine; intended for forensic and research applications
Catalog No. 50-297-0132 Supplier Cayman Chemical Supplier No. 143781 MG
An expected metabolite of BB-22 that would be detectable both in serum and urine; intended for forensic and research applications