Learn More
Cayman Chemical BB-22 3-hydroxyquinoline isom
SDP
Catalog No. 502970420
Encompass_Preferred
Differs from BB-22 structurally by having the hydroxyquinoline group attached at the three carbon instead of through the eight position of the ring; intended for forensic and research applications
Catalog No. 50-297-0420 Supplier Cayman Chemical Supplier No. 1450510 MG
Differs from BB-22 structurally by having the hydroxyquinoline group attached at the three carbon instead of through the eight position of the ring; intended for forensic and research applications