Learn More
Cayman Chemical BB-22 4-hydroxyisoquinoline isomer, Purity: ≥98%, CAS: 2365471-21-0, 10 mg
SDP
Catalog No. 502971010
Encompass_Preferred
Differs from BB-22 structurally by having an isoquinoline ring attached at the four position instead of a quinoline attached through the eight position of the ring; intended for forensic and research applications
Catalog No. 50-297-1010 Supplier Cayman Chemical Supplier No. 1454110 MG
Differs from BB-22 structurally by having an isoquinoline ring attached at the four position instead of a quinoline attached through the eight position of the ring; intended for forensic and research applications