Learn More
Cayman Chemical BB-22 4-hydroxyquinoline isomer, Purity: ≥98%, CAS: 2365470-95-5, 10 mg
SDP
Catalog No. 502969988
Encompass_Preferred
Differs from BB-22 structurally by having the hydroxyquinoline group attached at the four carbon instead of through the eight position of the ring; intended for forensic and research applications
Catalog No. 50-296-9988 Supplier Cayman Chemical Supplier No. 1450610 MG
Differs from BB-22 structurally by having the hydroxyquinoline group attached at the four carbon instead of through the eight position of the ring; intended for forensic and research applications