Learn More
Cayman Chemical BB-22 5-hydroxyisoquinoline isomer, Purity: ≥98%, CAS: 2365471-34-5, 10 mg
SDP
Catalog No. 502971074
Encompass_Preferred
Differs from BB-22 structurally by having an isoquinoline ring attached at the five carbon instead of a quinoline group attached through the eight position of the ring; intended for forensic and research applications
Catalog No. 50-297-1074 Supplier Cayman Chemical Supplier No. 1454210 MG
Differs from BB-22 structurally by having an isoquinoline ring attached at the five carbon instead of a quinoline group attached through the eight position of the ring; intended for forensic and research applications