Learn More
Cayman Chemical BB-22 5-hydroxyquinoline isomer, Purity: ≥98%, CAS: 2365471-40-3, 10 mg
SDP
Catalog No. 502971077
Encompass_Preferred
Differs from BB-22 structurally by having the hydroxyquinoline group attached at the five position instead of through the eight position of the ring; intended for forensic and research applications
Catalog No. 50-297-1077 Supplier Cayman Chemical Supplier No. 1450710 MG
Differs from BB-22 structurally by having the hydroxyquinoline group attached at the five position instead of through the eight position of the ring; intended for forensic and research applications